Learn More
American Research Products Inc Rat Vascular endothelial cell
Supplier:  American Research Products Inc CSBE07350R
Encompass
Rat Vascular endothelial cell growth factor receptor 1 VEGFR-1/Flt1 ELISA Kit from CUSABIO Detection Range 1 56 pg/mL-100 pg/mL Sensitivity 0 39 pg/mL Sample serum plasma tissue homogenates Method Sandwich
Catalog No. 50-003-26488