Learn More
American Research Products Inc Rat Vascular Endothelial cell
Catalog No. 5000328778
Encompass
Rat Vascular Endothelial cell Growth Factor VEGF ELISA Kit from CUSABIO Detection Range 3 9 pg/mL-250 pg/mL Sensitivity 0 97 pg/mL Sample serum plasma tissue homogenates Method Sandwich
Catalog No. 50-003-28778 Supplier American Research Products Inc Supplier No. CSBE04757R
Rat Vascular Endothelial cell Growth Factor VEGF ELISA Kit from CUSABIO Detection Range 3 9 pg/mL-250 pg/mL Sensitivity 0 97 pg/mL Sample serum plasma tissue homogenates Method Sandwich