Learn More
Biomatik Corporation Rat Vascular Endothelial Growth Factor A (VEGFA) CLIA Kit
Supplier:  Biomatik Corporation EKU08850
Encompass_Preferred
Rat Vascular Endothelial Growth Factor A (VEGFA) CLIA Kit for detecting VEGFA in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 2.74-2,000pg/ml. Sensitivity: 0.99pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Chemiluminescence. Assay Time: 2h, 40min. Accession Number: P16612. Gene ID: 83785. Target Synonyms: VEGF; VEGF-A; VPF; Vascular Endothelial Cell Growth Factor A; Vascular Permeability Factor. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-252-8613