Learn More
Biomatik Corporation Rat VEGF Co Regulated Chemokine 1 (VCC1) CLIA Kit
Supplier:  Biomatik Corporation EKU09878
Encompass_Preferred
Rat VEGF Co Regulated Chemokine 1 (VCC1) CLIA Kit for detecting VCC1 in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 1.37-1,000pg/ml. Sensitivity: 0.57pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Chemiluminescence. Assay Time: 2h, 40min. Accession Number: D4A875. Gene ID: 308436. Target Synonyms: CXCL17; SCYB17; DMC; Chemokine C-X-C-Motif Ligand 7; Dendritic cell and monocyte chemokine-like protein. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-5447