Learn More
Sino Biological Anti-VEGF164 Antibody, Rabbit Monoclonal
Supplier:  Sino Biological 80006R001100
Encompass_Preferred
This antibody was obtained from a rabbit immunized with purified, recombinant Rat VEGF164/VEGFA (rR VEGF/VEGFA; Catalog#80006-RNAB; AAA41211.1; Met1-Arg190).
Catalog No. 50-240-8965