Learn More
American Research Products Inc Rat Very low density lipoprote
Catalog No. 5000329347
Encompass
Rat Very low density lipoprotein (VLDL) ELISA Kit from CUSABIO Detection Range 3 12 ng/mL-200 ng/mL Sensitivity 0 78 ng/mL Sample serum plasma tissue homogenates Method Competitive
Catalog No. 50-003-29347 Supplier American Research Products Inc Supplier No. CSBE17088R
Rat Very low density lipoprotein (VLDL) ELISA Kit from CUSABIO Detection Range 3 12 ng/mL-200 ng/mL Sensitivity 0 78 ng/mL Sample serum plasma tissue homogenates Method Competitive