Learn More
Biomatik Corporation Rat Very Low Density Lipoprotein Receptor (VLDLR) ELISA Kit
Supplier:  Biomatik Corporation EKU08130
Encompass_Preferred
Rat Very Low Density Lipoprotein Receptor (VLDLR) ELISA Kit for detecting VLDLR in tissue homogenates, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 0.781-50ng/ml. Sensitivity: 0.31ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P98166. Target Synonyms: VLDL-R. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-6240