Learn More
BIOHIPPO Rat VGF Microsample
SDP
Catalog No. 5000313976
Encompass
Rat VGF Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of VGF in serum plasma tissue homogenates cell lysates cell culture supernates and other biological fluids It provides a detection range of 46 88-3000 pg/mL with a sensitivity of 19 72 pg/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Rat VGF Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Rat VGF Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Rat VGF biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color
Catalog No. 50-003-13976 Supplier BIOHIPPO Supplier No. BHE15211210
Rat VGF Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of VGF in serum plasma tissue homogenates cell lysates cell culture supernates and other biological fluids It provides a detection range of 46 88-3000 pg/mL with a sensitivity of 19 72 pg/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Rat VGF Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Rat VGF Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Rat VGF biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color