Learn More
American Research Products Inc Rat VGF NerVe Growth Factor In
Catalog No. 5000328519
Encompass
Rat VGF NerVe Growth Factor Inducible (VGF) ELISA Kit from CUSABIO Detection Range 0 312 ng/mL-20 ng/mL Sensitivity 0 078 ng/mL Sample serum plasma Method Sandwich
Catalog No. 50-003-28519 Supplier American Research Products Inc Supplier No. CSBE17520R
Rat VGF NerVe Growth Factor Inducible (VGF) ELISA Kit from CUSABIO Detection Range 0 312 ng/mL-20 ng/mL Sensitivity 0 078 ng/mL Sample serum plasma Method Sandwich