Learn More
BIOHIPPO Rat VIM Microsample
SDP
Catalog No. 5000315079
Encompass
Rat VIM Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of VIM in Serum plasma tissue homogenates and other biological fluids It provides a detection range of 0 32-20 ng/mL with a sensitivity of 0 128 ng/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Rat VIM Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Rat VIM Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Rat VIM biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color
Catalog No. 50-003-15079 Supplier BIOHIPPO Supplier No. BHE15210669
Rat VIM Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of VIM in Serum plasma tissue homogenates and other biological fluids It provides a detection range of 0 32-20 ng/mL with a sensitivity of 0 128 ng/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Rat VIM Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Rat VIM Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Rat VIM biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color