Learn More
BIOHIPPO Rat VIP Microsample
SDP
Catalog No. 5000315242
Encompass
Rat VIP Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of VIP in Serum plasma and other biological fluids It provides a detection range of 7 82-500 pg/mL with a sensitivity of 2 6 pg/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Rat VIP Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Rat VIP Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Rat VIP biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color
Catalog No. 50-003-15242 Supplier BIOHIPPO Supplier No. BHE15212234
Rat VIP Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of VIP in Serum plasma and other biological fluids It provides a detection range of 7 82-500 pg/mL with a sensitivity of 2 6 pg/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Rat VIP Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Rat VIP Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Rat VIP biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color