Learn More
Biomatik Corporation Rat Visfatin (VF) ELISA Kit
Supplier:  Biomatik Corporation EKU11521
Encompass_Preferred
Rat Visfatin (VF) ELISA Kit for detecting VF in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 1.23-100ng/ml. Sensitivity: 0.47ng/ml. Format: 96-Well Plate. Assay Type: Competitive Inhibition. Detection Method: Colormetric. Assay Time: 2h. Accession Number: Q80Z29. Gene ID: 297508. Target Synonyms: Nampt; NAmPRTase; PBEF; PBEF1; Pre-B-Cell Colony Enhancing Factor; Nicotinamide Phosphoribosyltransferase. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-252-8131