Learn More
Biomatik Corporation Rat Vitamin B12, Vb12 Elisa Kit
Supplier:  Biomatik Corporation EKC31503
Encompass_Preferred
Rat Vitamin B12, Vb12 Elisa Kit for detecting Vitamin B12, Vb12 in serum, plasma. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 0.312-20pg/ml. Sensitivity: 0.299pg/ml. Format: 96-Well Plate. Assay Type: Competitive. Assay Time: 1-5h. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-5293