Learn More
American Research Products Inc Rat Vitamin D3 receptor VDR
Catalog No. 5000328283
Encompass
Rat Vitamin D3 receptor (VDR) ELISA kit from CUSABIO Detection Range 31 2 pg/mL-2000 pg/mL Sensitivity 7 8 pg/mL Sample serum plasma tissue homogenates Method Sandwich
Catalog No. 50-003-28283 Supplier American Research Products Inc Supplier No. CSBEL025832RA
Rat Vitamin D3 receptor (VDR) ELISA kit from CUSABIO Detection Range 31 2 pg/mL-2000 pg/mL Sensitivity 7 8 pg/mL Sample serum plasma tissue homogenates Method Sandwich