Learn More
Biomatik Corporation Rat Von Hippel Lindau Tumor Suppressor (vHL) ELISA Kit
Supplier:  Biomatik Corporation EKU08182
Encompass_Preferred
Rat Von Hippel Lindau Tumor Suppressor (vHL) ELISA Kit for detecting vHL in tissue homogenates, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 0.312-20ng/ml. Sensitivity: 0.113ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q64259. Gene ID: 24874. Target Synonyms: HRCA1; RCA1; VHL1; Von Hippel-Lindau Syndrome. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-5017