Learn More
Sino Biological Anti-HE4/WFDC2 Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 80084T24100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Rat WAP5 / WFDC2 / HE4 isoform 2 (Q8CHN3-2; Met 4-Phe 120). WAP5 / WFDC2 / HE4 specific IgG was purified by Rat WAP5 / WFDC2 / HE4 affinity chromatography.
Catalog No. 50-255-2834