Learn More
Lonza Walkersville Rat, Wistar Hannover Hepatocytes, Cryopreserved, Suspension
Catalog No. NC1811016
Encompass
Cryopreserved Wistar Han Rat Hepatocytes, Suspension are specifically pre tested for metabolic activity of general Phase I and Phase II metabolism in suspension using ECOD outputs 7EC formation and formation of 7EC SULT and UGT products.This produce is not suitable for applications requiring adherant cells.Applications:Drug metabolism in suspension formatSpecies-species comparisonsGenomicsContent:Cryo vial, > 5 million viable cells
Catalog No. NC1811016 Supplier Lonza Walkersville Supplier No. RWCS01
Cryopreserved Wistar Han Rat Hepatocytes, Suspension are specifically pre tested for metabolic activity of general Phase I and Phase II metabolism in suspension using ECOD outputs 7EC formation and formation of 7EC SULT and UGT products.This produce is not suitable for applications requiring adherant cells.Applications:Drug metabolism in suspension formatSpecies-species comparisonsGenomicsContent:Cryo vial, > 5 million viable cells