Learn More
Lonza Walkersville Rat Wistar Hannover Hepatocytes, Cryopreserved, Plateable
Catalog No. NC2398987
Encompass
Cryopreserved Wistar Han Rat Hepatocytes, plateable qualified are specifically pre tested for metabolic activity of general Phase I and Phase II metabolism in suspension. They are tested for healthy morphology lasting 72 hours in monlayer sandwich culture.Benefits:Plateable>5 million cells/vialCharacterized for drug metabolismApplications:Drug metabolism in plated monolayer formatSpecies-species comparisonsGenomicsContent:Cryo vial, > 5 million viable cells
Catalog No. NC2398987 Supplier Lonza Walkersville Supplier No. RWCP01
Cryopreserved Wistar Han Rat Hepatocytes, plateable qualified are specifically pre tested for metabolic activity of general Phase I and Phase II metabolism in suspension. They are tested for healthy morphology lasting 72 hours in monlayer sandwich culture.Benefits:Plateable>5 million cells/vialCharacterized for drug metabolismApplications:Drug metabolism in plated monolayer formatSpecies-species comparisonsGenomicsContent:Cryo vial, > 5 million viable cells