Learn More
Lonza Walkersville Rat Wistar Heps Suspension
Catalog No. 5000496241
Encompass
All our suspension cryopreserved primary animal hepatocytes are specifically pre-tested for metabolic activity of general Phase I and Phase II metabolism in suspension They are not suitable for applications requiring adherent cells
Catalog No. 50-004-96241 Supplier Lonza Walkersville Supplier No. RICS01
All our suspension cryopreserved primary animal hepatocytes are specifically pre-tested for metabolic activity of general Phase I and Phase II metabolism in suspension They are not suitable for applications requiring adherent cells