Learn More
Braintree Scientific Rat ""Y"" Connector Catheter, 4.5F, 40mm L (sterile)
SDP
Catalog No. 501956164
Encompass
"Y" Connector Catheter is designed to be used with a syringe pump for infusion and withdrawal of fluids. Primarily used to adapt two osmotic pumps to one catheter line. Double outlet 0.039" od x 0.023 id. Single outlet 0.052" od x 0.034" id. PolyEthylene. **Sold for research purposes only and not for human use**
Catalog No. 50-195-6164 Supplier Braintree Scientific Supplier No. Y0225E0
"Y" Connector Catheter is designed to be used with a syringe pump for infusion and withdrawal of fluids. Primarily used to adapt two osmotic pumps to one catheter line. Double outlet 0.039" od x 0.023 id. Single outlet 0.052" od x 0.034" id. PolyEthylene. **Sold for research purposes only and not for human use**