Learn More
Biomatik Corporation Rat Zona Pellucida Glycoprotein 2, Sperm Receptor (ZP2) ELISA Kit
Supplier:  Biomatik Corporation EKU08255
Encompass_Preferred
Rat Zona Pellucida Glycoprotein 2, Sperm Receptor (ZP2) ELISA Kit for detecting ZP2 in tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 0.312-20ng/ml. Sensitivity: 0.125ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: O54767. Gene ID: 81828. Target Synonyms: ZPA; Zona Pellucida Sperm-Binding Protein 2 ; Zona pellucida protein A. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-6382