Learn More
MSC RATCH CLIMB VNTD HELMET
Catalog No. 5000781106
Encompass
The SecureFit safety helmet offers exceptional comfort and protection, with a self-adjusting suspension system for all-day comfort and superior protection.
  • Maintains comfortable pressure and protection.
  • Unique suspension with pressure diffusion for personalized fit.
  • Reduces forehead pressure for increased comfort.
  • Fits wide range of head sizes.
  • Optional venting for increased airflow.
  • Easy-turn ratchet suspension for secure fit.
  • Sits low on head for enhanced security.
  • UVicator sensor for UV exposure replacement cue.
  • Integrated clips for goggles and headlamps.
  • Six-point suspension for customizable height and even weight distribution.
  • Selectable chinstrap system for adaptable retention.
  • Meets ANSI Z89.1 Type I and EN12492 standards.
Catalog No. 50-007-81106 Supplier MSC Supplier No. 16987810
The SecureFit safety helmet offers exceptional comfort and protection, with a self-adjusting suspension system for all-day comfort and superior protection.
  • Maintains comfortable pressure and protection.
  • Unique suspension with pressure diffusion for personalized fit.
  • Reduces forehead pressure for increased comfort.
  • Fits wide range of head sizes.
  • Optional venting for increased airflow.
  • Easy-turn ratchet suspension for secure fit.
  • Sits low on head for enhanced security.
  • UVicator sensor for UV exposure replacement cue.
  • Integrated clips for goggles and headlamps.
  • Six-point suspension for customizable height and even weight distribution.
  • Selectable chinstrap system for adaptable retention.
  • Meets ANSI Z89.1 Type I and EN12492 standards.