Learn More
MSC RATCH CLIMB VNTD HELMET
Catalog No. 5000767650
Encompass
The 3M™ SecureFit™ X5000 Safety Helmet is a vented, Class C certified hard hat offering superior comfort and protection. It features 3M™ SecureFit™ Pressure Diffusion Technology and a 6-point ratchet suspension system.
  • SecureFit Pressure Diffusion Technology reduces forehead pressure by 20% and a 6-point ratchet suspension provides a secure, customizable fit.
  • Vented shell ensures enhanced airflow and comfort.
  • UVicator sensor signals helmet replacement due to UV exposure.
  • Integrated clips for securing goggles and headlamps.
  • Premium chin strap with magnetic quick-release buckle, selectable for EN397 or EN12492 retention.
  • Highly reflective material for improved visibility and durability.
  • Constructed from UV stabilized ABS, nylon, HDPE, and coated foam.
  • Meets ANSI Z89.1 Type I, Class C requirements.
Catalog No. 50-007-67650 Supplier MSC Supplier No. 16987877
The 3M™ SecureFit™ X5000 Safety Helmet is a vented, Class C certified hard hat offering superior comfort and protection. It features 3M™ SecureFit™ Pressure Diffusion Technology and a 6-point ratchet suspension system.
  • SecureFit Pressure Diffusion Technology reduces forehead pressure by 20% and a 6-point ratchet suspension provides a secure, customizable fit.
  • Vented shell ensures enhanced airflow and comfort.
  • UVicator sensor signals helmet replacement due to UV exposure.
  • Integrated clips for securing goggles and headlamps.
  • Premium chin strap with magnetic quick-release buckle, selectable for EN397 or EN12492 retention.
  • Highly reflective material for improved visibility and durability.
  • Constructed from UV stabilized ABS, nylon, HDPE, and coated foam.
  • Meets ANSI Z89.1 Type I, Class C requirements.