Learn More
MSC RATCH CLIMB VTND HELMET
Catalog No. 5000677158
Encompass
This hard hat provides head protection with its durable Acrylonitrile Butadiene Styrene (ABS) construction and comfortable 6-point suspension system.
  • Ratchet adjustment for a secure fit
  • Vented for improved airflow
  • Slotted for accessory attachment
  • Universal head size fit
  • ANSI Type I compliant
Catalog No. 50-006-77158 Supplier MSC Supplier No. 16987869
This hard hat provides head protection with its durable Acrylonitrile Butadiene Styrene (ABS) construction and comfortable 6-point suspension system.
  • Ratchet adjustment for a secure fit
  • Vented for improved airflow
  • Slotted for accessory attachment
  • Universal head size fit
  • ANSI Type I compliant