Learn More
MSC RATCH CLIMBING HELMET
Catalog No. 5000676860
Encompass
3M™ SecureFit™ X5000 Series Safety Helmets provide comfort and security with a climbing helmet-inspired design. They meet occupational safety requirements and feature exclusive 3M™ Pressure Diffusion Technology in the suspension system.
  • Suspension system with exclusive Pressure Diffusion Technology
  • Compatible with various faceshields, earmuffs, and other accessories
  • Meets ANSI Z89.1 standards
  • Climbing helmet-inspired design with customizable fit
  • Features a 4-point chin strap with buckle and selector, switching between EN 12492 and EN 397 standards
  • Features accessory slots, replaceable brow pad, and UV indicator
  • Type I hard hat with ABS shell material
  • Includes a 6-point ratchet suspension, available in vented options
Catalog No. 50-006-76860 Supplier MSC Supplier No. 16987950
3M™ SecureFit™ X5000 Series Safety Helmets provide comfort and security with a climbing helmet-inspired design. They meet occupational safety requirements and feature exclusive 3M™ Pressure Diffusion Technology in the suspension system.
  • Suspension system with exclusive Pressure Diffusion Technology
  • Compatible with various faceshields, earmuffs, and other accessories
  • Meets ANSI Z89.1 standards
  • Climbing helmet-inspired design with customizable fit
  • Features a 4-point chin strap with buckle and selector, switching between EN 12492 and EN 397 standards
  • Features accessory slots, replaceable brow pad, and UV indicator
  • Type I hard hat with ABS shell material
  • Includes a 6-point ratchet suspension, available in vented options