Learn More
MSC RATCH CLIMBING HELMET
Catalog No. 5000677900
Encompass
The 3M Hard Hat features a 6-point suspension system, providing reliable head protection. Made from durable ABS material, this slotted hard hat is suitable for various industrial applications.
  • Standard type
  • White color
  • Ratchet adjustment type
  • ABS material
  • ANSI Type I
  • Not vented
  • Slotted design
  • Universal head size fit
  • Six-point suspension
  • Country of origin: Poland
Catalog No. 50-006-77900 Supplier MSC Supplier No. 16987836
The 3M Hard Hat features a 6-point suspension system, providing reliable head protection. Made from durable ABS material, this slotted hard hat is suitable for various industrial applications.
  • Standard type
  • White color
  • Ratchet adjustment type
  • ABS material
  • ANSI Type I
  • Not vented
  • Slotted design
  • Universal head size fit
  • Six-point suspension
  • Country of origin: Poland