Learn More
Grainger Condor Ratchet Faceshield Assembly, Clear Polycarbonate Visor, Uncoated, 1 ea
Catalog No. NC2152186
Encompass_Preferred
This ratchet faceshield assembly provides reliable face protection with a clear polycarbonate visor. Designed for versatility and comfort, the unit features a partial crown headgear with a convenient ratchet suspension system for secure and adjustable fitting. It is suitable for a range of professional and industrial safety applications.
  • Features a spark deflector with a wide range of adjustability
  • Constructed with a durable polycarbonate visor and plastic headgear
  • Includes a user-friendly ratchet suspension system
  • Meets ANSI Z87.1-2010 standards
  • Designed for reusable applications
  • Features an uncoated clear visor
This item is not returnable. View return policy
Catalog No. NC2152186 Supplier Grainger Supplier No. 4EZC2
This ratchet faceshield assembly provides reliable face protection with a clear polycarbonate visor. Designed for versatility and comfort, the unit features a partial crown headgear with a convenient ratchet suspension system for secure and adjustable fitting. It is suitable for a range of professional and industrial safety applications.
  • Features a spark deflector with a wide range of adjustability
  • Constructed with a durable polycarbonate visor and plastic headgear
  • Includes a user-friendly ratchet suspension system
  • Meets ANSI Z87.1-2010 standards
  • Designed for reusable applications
  • Features an uncoated clear visor