Learn More
Glass Expansion Ratchet Connector
SDP
Supplier:  Glass Expansion 708031350
Encompass_Preferred
Ratchet Connector. Glass Expansion’s ¼-28 Ratchet Connector Fitting for making capillary connections to ICP-OES and ICP-MS sample introduction components. Can be used with 1.6mm OD (1/16”) ferrules P/N 70-803-0749 or with 1.3mm OD ferrules P/N 70-803-0748 to terminate capillary tubing where ratchet fitting is required. Swage Tool for Ratchet Connector, P/N 50-100-1811, secures ferrule on capillary and allows desired capillary tube length to be terminated with appropriate ferrule and ratchet fitting.Glass Expansion is ISO 9001:2015 certified by AB Certification, No. AU4501-9001
Catalog No. 50-270-3060