Learn More
Glass Expansion Ratchet EzyLok Assembly
SDP
Catalog No. 502703053
Encompass_Preferred
Ratchet EzyLok Assembly. Compatible with Glass Expansion VeeSpray Ceramic DC Nebulizers for Agilent 5000 Series and Thermo 6000/7000 Duo, Radial, PRO Duo, PRO Radial Series ICP-OES instruments (501907391 and 501694634; A13-07-CV6 and A31-07-CV6) to connect argon gas line directly to instrument argon supply.Glass Expansion is ISO 9001:2015 certified by AB Certification, No. AU4501-9001
Catalog No. 50-270-3053 Supplier Glass Expansion Supplier No. 708031485
Ratchet EzyLok Assembly. Compatible with Glass Expansion VeeSpray Ceramic DC Nebulizers for Agilent 5000 Series and Thermo 6000/7000 Duo, Radial, PRO Duo, PRO Radial Series ICP-OES instruments (501907391 and 501694634; A13-07-CV6 and A31-07-CV6) to connect argon gas line directly to instrument argon supply.Glass Expansion is ISO 9001:2015 certified by AB Certification, No. AU4501-9001