Learn More
Grainger Proto Ratchet Repair Kit, 1/4 in. Drive, Pear Head, 10-Piece Set
Catalog No. 502793342
Encompass_Preferred
This repair kit is designed to restore 1/4 in. drive pear head ratchets to optimal working condition. The 10-piece set includes all necessary components to service and maintain compatible ratchet handles, ensuring consistent performance in demanding applications.
  • Includes springs, screws, a 24-tooth gear plate, and additional replacement parts
  • Designed for 1/4 in. drive pear head ratchets
  • Compatible with models J4749 and J4749BL
  • Constructed from durable steel
  • Meets ASME B107.110-2012 and ASME B107.500 standards
Catalog No. 50-279-3342 Supplier Grainger Supplier No. 1AF40
This repair kit is designed to restore 1/4 in. drive pear head ratchets to optimal working condition. The 10-piece set includes all necessary components to service and maintain compatible ratchet handles, ensuring consistent performance in demanding applications.
  • Includes springs, screws, a 24-tooth gear plate, and additional replacement parts
  • Designed for 1/4 in. drive pear head ratchets
  • Compatible with models J4749 and J4749BL
  • Constructed from durable steel
  • Meets ASME B107.110-2012 and ASME B107.500 standards