Learn More
Grainger Snap-Loc Ratchet Strap, Polyester Webbing, 8 ft., 4,400 lb. Break Strength, 1 ea.
Catalog No. 502796011
Encompass_Preferred
This heavy-duty 8 ft. E-Track ratchet strap is designed for secure cargo control in trucks, trailers, vans, and other transport applications. Constructed from durable 2 in. wide polyester webbing, the strap features a 4,400 lb. break strength and a 1,467 lb. working load limit to safely bundle and tie down items such as pipe, lumber, and ladders. The system utilizes zinc-plated steel hardware to resist weather and environmental contaminants while providing a secure lock that prevents accidental disengagement during load shifts.
  • USA made with durable 2 in. wide polyester webbing
  • 4,400 lb. assembly break strength and 1,467 lb. working load limit
  • Zinc-plated steel hardware for corrosion resistance
  • Includes hook and loop storage fastener for neat organization
  • Spring-loaded lever for quick connection and disconnection
  • Compatible with standard E-Track systems
Catalog No. 50-279-6011 Supplier Grainger Supplier No. 19XZ69
This heavy-duty 8 ft. E-Track ratchet strap is designed for secure cargo control in trucks, trailers, vans, and other transport applications. Constructed from durable 2 in. wide polyester webbing, the strap features a 4,400 lb. break strength and a 1,467 lb. working load limit to safely bundle and tie down items such as pipe, lumber, and ladders. The system utilizes zinc-plated steel hardware to resist weather and environmental contaminants while providing a secure lock that prevents accidental disengagement during load shifts.
  • USA made with durable 2 in. wide polyester webbing
  • 4,400 lb. assembly break strength and 1,467 lb. working load limit
  • Zinc-plated steel hardware for corrosion resistance
  • Includes hook and loop storage fastener for neat organization
  • Spring-loaded lever for quick connection and disconnection
  • Compatible with standard E-Track systems