Learn More
BioVendor, LLC RATLAPS CTX-I EIA (96 wells)
SDP
Catalog No. 501963837
Encompass
Competitive ELISA, Immobilized antigenRatLaps™ (CTX-I) EIA is an enzyme-linked immunosorbent assay for the quantitative determination of bone-related degradation products from C-terminal telopeptides of type I collagen in rat/mouse serum or urine, and from rat/mouse bone released into cell culture supernatants by osteoclasts.The RatLaps™ (CTX-I) EIA is based on the observation that certain C-telopeptide degradation products from type I collagen released during osteoclastic bone resorption. ApplicationsSerum, Urine, Plasma, Cell culture supernatantSample Requirements20 μL/wellCalibration Range0-200 ng/mLLimit of Detection2.0 ng/mL
Catalog No. 50-196-3837 Supplier BioVendor, LLC Supplier No. AC06F1
Competitive ELISA, Immobilized antigenRatLaps™ (CTX-I) EIA is an enzyme-linked immunosorbent assay for the quantitative determination of bone-related degradation products from C-terminal telopeptides of type I collagen in rat/mouse serum or urine, and from rat/mouse bone released into cell culture supernatants by osteoclasts.The RatLaps™ (CTX-I) EIA is based on the observation that certain C-telopeptide degradation products from type I collagen released during osteoclastic bone resorption. ApplicationsSerum, Urine, Plasma, Cell culture supernatantSample Requirements20 μL/wellCalibration Range0-200 ng/mLLimit of Detection2.0 ng/mL