Learn More
SOMNI Scientific™ RatTaxic™ Stereotaxic Nose Cone For Active Waste Gas Scavenging
SDP
Catalog No. 04777142 Shop All Somni Scientific Products
Change view
Click to view available options
Includes:
Supply tubing,connectors
Supply tubing,connectors,head clamp
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Includes
04-777-142 Supply tubing,connectors,head clamp
04-777-140 Supply tubing,connectors
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 04-777-142 Supplier SOMNI Scientific™ Supplier No. NC19102
Add to Cart
Add to Cart
Designed to be used with stereotaxic devices to safely deliver and actively scavenge inhalant anesthesia.
  • Rat size
  • Stereotaxic nosecone
  • Use with attenuated active scavenging
  • Supply tubing and connectors included
  • Contact SOMNI Scientific to check compatibility
  • Works with SOMNI EPS-3 or attenuated in-house vacuum system
Specifications
For Use With (Application) Research Animal Inhalant Anesthesia
For Use With (Equipment) AMD-17106, AMD-17004, TT-17000, TT-17001, TT-17002, WG-15001, WG-15101
Material 3D Printed
Length (Metric) 6.35 cm
Width (Metric) 3 cm
Includes Supply tubing,connectors,head clamp
For Veterinary Use Only