Learn More
Zymo Research Corporation Rattler™ Plating Beads (230 g) (5 Bottles) ^
SDP
Supplier:  Zymo Research Corporation S10015
Encompass
The sterile glass beads are simply poured onto solid plated medium together with a liquid cell suspension, and the mixture is shaken to distribute the cells evenly over the medium's surface. This allows for numerous plates to be processed quickly and efficiently. Pour the Rattler Beads onto a series of plates, stack, and shake simultaneously in a side-to-side motion. The beads can be easily removed following inversion of the plates and pouring off from the plate lids. Using the Rattler Plating Beads is simple, easy, and saves you time. The beads come sterile in polycarbonate bottles and can be reused following cleaning and washing.Rattler™ Plating Beads Fast & Easy-To-Use: Simply pour beads onto the plate, and shake to quickly spread cells evenly over the entire surface. Ideal for processing multiple plates simultaneously. No Flaming Required: 4.5 mm glass plating beads are provided sterile in polycarbonate bottles. Reusable: Just clean and autoclave after use.
Catalog No. 50-444-635