Learn More
Selleck Chemical LLC Rauwolscine hydrochloride S5337-25mg
SDP
Catalog No. 503108557
Encompass_Preferred
Rauwolscine hydrochloride (Isoyohimbine -Yohimbine corynanthidine) is the hydrochloride salt form of Rauwolscine a specific and potent 2 antagonist with Ki of 12 nM
Catalog No. 50-310-8557 Supplier Selleck Chemical LLC Supplier No. S533725MG
Rauwolscine hydrochloride (Isoyohimbine -Yohimbine corynanthidine) is the hydrochloride salt form of Rauwolscine a specific and potent 2 antagonist with Ki of 12 nM