Learn More
Cayman Chemical Rauwolscine (hydrochloride), Purity: ≥98%, CAS: 6211-32-1, 100 mg
SDP
Catalog No. 502969197
Encompass_Preferred
A natural alkaloid that acts as a selective and reversible α2-adrenergic receptor antagonist (Ki = 12 nM); also acts as a receptor antagonist at the serotonin 5-HT2B receptor (Ki = 14.3 nM)
Catalog No. 50-296-9197 Supplier Cayman Chemical Supplier No. 15596100 MG
A natural alkaloid that acts as a selective and reversible α2-adrenergic receptor antagonist (Ki = 12 nM); also acts as a receptor antagonist at the serotonin 5-HT2B receptor (Ki = 14.3 nM)