Learn More
Cayman Chemical RauwolscInhydrochlorIde 50mg
SDP
Catalog No. 502971381
Encompass_Preferred
A natural alkaloid that acts as a selective and reversible α2-adrenergic receptor antagonist (Ki = 12 nM); also acts as a receptor antagonist at the serotonin 5-HT2B receptor (Ki = 14.3 nM)
Catalog No. 50-297-1381 Supplier Cayman Chemical Supplier No. 1559650 MG
A natural alkaloid that acts as a selective and reversible α2-adrenergic receptor antagonist (Ki = 12 nM); also acts as a receptor antagonist at the serotonin 5-HT2B receptor (Ki = 14.3 nM)