Learn More
Raven Environmental Products Raven F-10300 Horizontal Process Centrifuge
Catalog No. NC9817581
Encompass_Preferred
The Raven F-10300 horizontal process centrifuge is specifically designed for sampling applications to determine solids concentration in percent volume as low as two tenths of one percent (0.2%.) Keep track of solids in aeration tanks, clarifiers, waste sludge lines, return sludge lines, digesters and more. You'll get answers when you need them and verify your adjustments in real time. Six place horizontal rotor included. Tube capacity up to 15ml with optional rotor buckets. Electronic timer from 1 to 999 minutes. A bright green LED alerts the operator when the unit is powered on. The LCD display indicates when the rotor is spinning. An audible sounds when the cycle is complete. 3000 RPM with 1,792 relative centifugal force (RCF). 17 in. wide and 13 in. tall. Stainless steel and aluminum structure for durability in harsh environments such as industrial and wastewater facilities.
Catalog No. NC9817581 Supplier Raven Environmental Products Supplier No. F10300
The Raven F-10300 horizontal process centrifuge is specifically designed for sampling applications to determine solids concentration in percent volume as low as two tenths of one percent (0.2%.) Keep track of solids in aeration tanks, clarifiers, waste sludge lines, return sludge lines, digesters and more. You'll get answers when you need them and verify your adjustments in real time. Six place horizontal rotor included. Tube capacity up to 15ml with optional rotor buckets. Electronic timer from 1 to 999 minutes. A bright green LED alerts the operator when the unit is powered on. The LCD display indicates when the rotor is spinning. An audible sounds when the cycle is complete. 3000 RPM with 1,792 relative centifugal force (RCF). 17 in. wide and 13 in. tall. Stainless steel and aluminum structure for durability in harsh environments such as industrial and wastewater facilities.