Learn More
Aviva Systems Biology Corp RAVER1 Antibody - N-terminal region (ARP41099_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP41099_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against RAVER1. It was validated on Western Blot and immunohistochemistry. The immunogen is a synthetic peptide directed towards the N terminal region of human RAVER1. Alternate names: ApplicationIHC, WB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Rabbit
Catalog No. 50-014-53271