Learn More
Aviva Systems Biology Corp RAVER1 Antibody - middle region (ARP41098_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP41098_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against RAVER1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human RAVER1. Alternate names: KIAA1978ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat
Catalog No. 50-014-57722