Learn More
Aviva Systems Biology Corp RAVER2 Antibody - middle region (ARP40904_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP40904_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against RAVER2. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human RAVER2. Alternate names: DKFZp762D1011, FLJ10770, KIAA1579ApplicationWB. Species reactivity: Cow, Human, Mouse, Rabbit
Catalog No. 50-014-57658