Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Ravidasvir hydrochloride | 1303533-81-4 | 99.9% | 835.82 g·mol⁻1 | C42H52Cl2N8O6 | 100MG
SDP

Catalog No. 5000238446
Encompass_Preferred

Ravidasvir hydrochloride is a research-grade, pan-genotypic inhibitor of hepatitis C virus (HCV) nonstructural protein 5A (NS5A), supplied as a white to off-white solid and used in antiviral in vitro and preclinical studies.

  • Pan-genotypic NS5A inhibitor with low-nanomolar EC50 against multiple HCV genotypes.
  • High purity: 99.9% (reported by supplier).
  • Molecular weight: 835.82 g·mol⁻1; formula C42H52Cl2N8O6.
  • Appearance: white to off-white solid, suitable for analytical and biological assays.
  • Storage: store sealed at 4°C; in solvent, -80°C for up to 6 months or -20°C for 1 month.
  • Intended for research use only; not for human or clinical use.

Catalog No. 50-002-38446 Supplier Medchemexpress LLC Supplier No. HY124191100MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Ravidasvir hydrochloride is a research-grade, pan-genotypic inhibitor of hepatitis C virus (HCV) nonstructural protein 5A (NS5A), supplied as a white to off-white solid and used in antiviral in vitro and preclinical studies.

  • Pan-genotypic NS5A inhibitor with low-nanomolar EC50 against multiple HCV genotypes.
  • High purity: 99.9% (reported by supplier).
  • Molecular weight: 835.82 g·mol⁻1; formula C42H52Cl2N8O6.
  • Appearance: white to off-white solid, suitable for analytical and biological assays.
  • Storage: store sealed at 4°C; in solvent, -80°C for up to 6 months or -20°C for 1 month.
  • Intended for research use only; not for human or clinical use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.