Learn More
Medchemexpress LLC Ravidasvir hydrochloride | 1303533-81-4 | 99.9% | 835.82 g/mol | C42H52Cl2N8O6 | 10MG
SDP
Catalog No. 5000238443
Encompass_Preferred
Ravidasvir hydrochloride is a research-grade small-molecule inhibitor of the hepatitis C virus NS5A protein, supplied as the hydrochloride salt for laboratory studies. It exhibits potent, pan-genotypic antiviral activity and is provided with high purity and supporting documentation for use in biochemical and cellular assays.
  • Pan-genotypic NS5A inhibitor with potent antiviral activity.
  • High purity (99.9%) suitable for research applications.
  • Supplied as a hydrochloride salt for improved solubility handling.
  • Recommended storage conditions provided to preserve stability.
  • Includes product datasheet and safety data sheet for lab use.
Catalog No. 50-002-38443 Supplier Medchemexpress LLC Supplier No. HY12419110MG
Ravidasvir hydrochloride is a research-grade small-molecule inhibitor of the hepatitis C virus NS5A protein, supplied as the hydrochloride salt for laboratory studies. It exhibits potent, pan-genotypic antiviral activity and is provided with high purity and supporting documentation for use in biochemical and cellular assays.
  • Pan-genotypic NS5A inhibitor with potent antiviral activity.
  • High purity (99.9%) suitable for research applications.
  • Supplied as a hydrochloride salt for improved solubility handling.
  • Recommended storage conditions provided to preserve stability.
  • Includes product datasheet and safety data sheet for lab use.