Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Ravidasvir hydrochloride | 1303533-81-4 | 99.9% | 835.82 g/mol | C42H52Cl2N8O6 | 1MG
SDP

Catalog No. 5000236040
Encompass_Preferred

Ravidasvir hydrochloride is a research-grade, pan-genotypic inhibitor of hepatitis C virus non-structural protein 5A (NS5A), supplied as a high-purity solid for in vitro and preclinical research. It is not for human or veterinary use and should be stored sealed and protected from moisture.

  • Pan-genotypic NS5A inhibitor suitable for HCV research.
  • High purity: 99.9%.
  • Molecular weight: 835.82 g/mol.
  • Chemical formula: C42H52Cl2N8O6.
  • Solid form; store sealed and away from moisture.
  • Intended for in vitro and preclinical studies only.

Catalog No. 50-002-36040 Supplier Medchemexpress LLC Supplier No. HY1241911MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Ravidasvir hydrochloride is a research-grade, pan-genotypic inhibitor of hepatitis C virus non-structural protein 5A (NS5A), supplied as a high-purity solid for in vitro and preclinical research. It is not for human or veterinary use and should be stored sealed and protected from moisture.

  • Pan-genotypic NS5A inhibitor suitable for HCV research.
  • High purity: 99.9%.
  • Molecular weight: 835.82 g/mol.
  • Chemical formula: C42H52Cl2N8O6.
  • Solid form; store sealed and away from moisture.
  • Intended for in vitro and preclinical studies only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.