Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Ravidasvir (hydrochloride) | 1303533-81-4 | 99.9% | 835.82 g/mol | C42H52Cl2N8O6 | 10MM 1ML
SDP

Catalog No. 5000236085
Encompass_Preferred

Ravidasvir hydrochloride is a pan-genotypic hepatitis C virus (HCV) NS5A inhibitor supplied for research use. It is provided as a ready-to-use 10 mM solution in DMSO (1 mL) and is intended for in vitro antiviral assays, medicinal chemistry, and analytical studies. The material is characterized by high purity and is supported by analytical documentation to aid reproducibility and assay development.

  • Pan-genotypic HCV NS5A inhibitor for antiviral research.
  • Provided as a 10 mM solution in DMSO, 1 mL vial.
  • High purity suitable for biological and analytical assays.
  • Characterized by supporting analytical documentation such as SDS and COA.
  • Molecular weight 835.82 g/mol; chemical formula C42H52Cl2N8O6.

Catalog No. 50-002-36085 Supplier Medchemexpress LLC Supplier No. HY12419110MM 1ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Ravidasvir hydrochloride is a pan-genotypic hepatitis C virus (HCV) NS5A inhibitor supplied for research use. It is provided as a ready-to-use 10 mM solution in DMSO (1 mL) and is intended for in vitro antiviral assays, medicinal chemistry, and analytical studies. The material is characterized by high purity and is supported by analytical documentation to aid reproducibility and assay development.

  • Pan-genotypic HCV NS5A inhibitor for antiviral research.
  • Provided as a 10 mM solution in DMSO, 1 mL vial.
  • High purity suitable for biological and analytical assays.
  • Characterized by supporting analytical documentation such as SDS and COA.
  • Molecular weight 835.82 g/mol; chemical formula C42H52Cl2N8O6.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.