Learn More
Medchemexpress LLC Ravidasvir hydrochloride | 1303533-81-4 | 99.9% | 835.82 g/mol | C42H52Cl2N8O6 | 25MG
SDP
Catalog No. 5000237006
Encompass_Preferred
Ravidasvir hydrochloride is a research-grade hepatitis C virus NS5A inhibitor used in preclinical and biochemical studies. It is supplied as a white to off-white solid and is available in solid and solution pack sizes. The product has a reported high purity (~99.9%), molecular formula C42H52Cl2N8O6, and molecular weight 835.82 g/mol. Storage recommendations include sealed storage at 4°C and long-term storage in solvent at -80°C.
Catalog No. 50-002-37006 Supplier Medchemexpress LLC Supplier No. HY12419125MG
Ravidasvir hydrochloride is a research-grade hepatitis C virus NS5A inhibitor used in preclinical and biochemical studies. It is supplied as a white to off-white solid and is available in solid and solution pack sizes. The product has a reported high purity (~99.9%), molecular formula C42H52Cl2N8O6, and molecular weight 835.82 g/mol. Storage recommendations include sealed storage at 4°C and long-term storage in solvent at -80°C.