Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Ravidasvir hydrochloride | 1303533-81-4 | MFCD32705191 | 99.9% | 835.82 | C42H52Cl2N8O6 | 50MG
SDP

Catalog No. 5000238442
Encompass_Preferred

Ravidasvir hydrochloride is the hydrochloride salt of ravidasvir, a pan-genotypic hepatitis C virus (HCV) NS5A inhibitor supplied as a high-purity research compound in solid form. It inhibits HCV replication at low nanomolar concentrations and is provided for laboratory research applications.

  • High purity (99.9%).
  • White to off-white solid physical form.
  • Molecular weight 835.82 g/mol.
  • Packaged as 50 MG for research use.
  • Storage: sealed at 4°C; in solvent: -80°C (up to 6 months) or -20°C (up to 1 month).
  • Provides potent pan-genotypic NS5A inhibition with low nanomolar activity.

Catalog No. 50-002-38442 Supplier Medchemexpress LLC Supplier No. HY12419150MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Ravidasvir hydrochloride is the hydrochloride salt of ravidasvir, a pan-genotypic hepatitis C virus (HCV) NS5A inhibitor supplied as a high-purity research compound in solid form. It inhibits HCV replication at low nanomolar concentrations and is provided for laboratory research applications.

  • High purity (99.9%).
  • White to off-white solid physical form.
  • Molecular weight 835.82 g/mol.
  • Packaged as 50 MG for research use.
  • Storage: sealed at 4°C; in solvent: -80°C (up to 6 months) or -20°C (up to 1 month).
  • Provides potent pan-genotypic NS5A inhibition with low nanomolar activity.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.