Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Ravidasvir (hydrochloride) | 1303533-81-4 | MFCD32705191 | 99.9% | 835.82 | C42H52Cl2N8O6 | 5MG
SDP

Catalog No. 5000236087
Encompass_Preferred

Ravidasvir hydrochloride is the hydrochloride salt of ravidasvir, a pan-genotypic hepatitis C virus (HCV) NS5A inhibitor supplied for research use. It is provided as solid vials in multiple sizes and as DMSO solutions for immediate assay use. Intended for in vitro research and assay development; not for human or clinical use.

  • High purity suitable for analytical and biological research.
  • Multiple solid vial sizes available, including small research quantities.
  • Available as a ready-to-use 10 mM solution in DMSO for rapid testing.
  • Molecular weight 835.82 and formula C42H52Cl2N8O6 for precise calculations.
  • CAS number 1303533-81-4 for unambiguous identification.

Catalog No. 50-002-36087 Supplier Medchemexpress LLC Supplier No. HY1241915MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Ravidasvir hydrochloride is the hydrochloride salt of ravidasvir, a pan-genotypic hepatitis C virus (HCV) NS5A inhibitor supplied for research use. It is provided as solid vials in multiple sizes and as DMSO solutions for immediate assay use. Intended for in vitro research and assay development; not for human or clinical use.

  • High purity suitable for analytical and biological research.
  • Multiple solid vial sizes available, including small research quantities.
  • Available as a ready-to-use 10 mM solution in DMSO for rapid testing.
  • Molecular weight 835.82 and formula C42H52Cl2N8O6 for precise calculations.
  • CAS number 1303533-81-4 for unambiguous identification.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.