Learn More
Cayman Chemical Ravidasvir (hydrochloride), Purity: ≥98%, CAS: 1303533-81-4, 10 mg
SDP
Catalog No. 502916710
Encompass_Preferred
An NS5A inhibitor; inhibits HCV replication in Huh7 replicon cells containing NS5A from the HCV genotypes 1a, -1b, and 3a (EC50s = 0.12, 0.01, and 1.14 nM, respectively)
Catalog No. 50-291-6710 Supplier Cayman Chemical Supplier No. 3779410 MG
An NS5A inhibitor; inhibits HCV replication in Huh7 replicon cells containing NS5A from the HCV genotypes 1a, -1b, and 3a (EC50s = 0.12, 0.01, and 1.14 nM, respectively)