Learn More
Medchemexpress LLC HY-15947A 5mg Medchemexpress, Ravoxertinib hydrochloride CAS:2070009-58-2 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY15947A5MG
Encompass_Preferred
Medchemexpress, HY-15947A 5mg Ravoxertinib HCl CAS:2070009-58-2 Ravoxertinib hydrochloride (GDC-0994 hydrochloride) is an orally bioavailable inhibitor selective for ERK kinase activity with IC50 of 6.1 nM and 3.1 nM for ERK1 and ERK2, respectively. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-9964